Få mere at vide
Tocris Bioscience™ Cytosporone B
High affinity Nur77 agonist
Brand: Tocris Bioscience™ 5459/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Cytosporone B is a naturally occurring Nur77 (NR4A1) agonist (Kd = 8.52 x 10-7 M). Induces apoptosis in gastric cancer cells in vitro. Increases blood glucose levels in fasting C57 mice and suppresses xenograft tumor growth. Also exhibits a potent antifibrotic effect in bleomycin-induced skin fibrosis in vivo.Tekniske data
| Ethyl 3,5-dihydroxy-2-(1-oxooctyl)-benzeneacetate | |
| 321661-62-5 | |
| 10 mg |