missing translation for 'onlineSavingsMsg'
Få mere at vide

Cytomegalovirus p65 Antibody (CMV100), Alexa Fluor™ 350, Novus Biologicals™

Artikelnummer. 18168404 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
18168404 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18168404 Leverandør Novus Biologicals Leverandørnr. NBP234595AF350

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Cytomegalovirus p65 Monoclonal specifically detects Cytomegalovirus p65 in Cytomegalovirus, Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Cytomegalovirus p65
Applikationer ELISA, Immunocytochemistry, Immunofluorescence
Klassifikation Monoclonal
Klon CMV100
Konjugeret Alexa Fluor 350
Fortynding ELISA, Immunocytochemistry/Immunofluorescence
Formulering 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CMV, p65
Værtsarter Mouse
Immunogen Recombinant CMV-p65 protein
Oprensningsmetode Protein A or G purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primær eller sekundær Primary
Testspecificitet Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.
Målarter Cytomegalovirus (CMV), Virus
Indhold og opbevaring Store at 4°C in the dark.
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.