Få mere at vide
Cytochrome C, TRC
Artikelnummer. 30362129
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362129 1 g
30365488 250 mg
30357884 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362129 Leverandør Toronto Research Chemicals Leverandørnr. TRCC2720151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 9007-43-6
Molekylær formel C42H52FeN8O6S2
Molekylvægt (g/mol) 884.89
Synonym Cromoci, CYC, CYCS, Cytochrome CymA, Cytomac P, Cytorest, Ferricytochrome c, Ferrocytochrome c
IUPAC navn iron(II) 2,17-bis(1-((2-amino-3-(methylamino)-3-oxopropyl)thio)ethyl)-8,12-bis(2-carboxyethyl)-3,7,13,18-tetramethylporphyrin-21,23-diide
SMIL CC1=C(C(C)SCC(N)C(NC)=O)C(/C=C(C(C)=C/2CCC(O)=O)\[N-]C2=C/C(C(CCC(O)=O)=C/3C)=NC3=C/4)=N/C1=C\C5=C(C(C)SCC(N)C(NC)=O)C(C)=C4[N-]5.[Fe+2]
Tekniske data
Kemisk navn eller materiale Cytochrome C
Mængde 1 g
InChI formel InChI=1S/C42H54N8O6S2.Fe/c1-19-25(9-11-37(51)52)33-16-34-26(10-12-38(53)54)20(2)30(48-34)14-35-40(24(6)58-18-28(44)42(56)46-8)22(4)32(50-35)15-36-39(21(3)31(49-36)13-29(19)47-33)23(5)57-17-27(43)41(55)45-7;/h13-16,23-24,27-28H,9-12,17-18,43-44H2,1-8H3,(H6,45,46,47,48,49,50,51,52,53,54,55,56);/q;+2/p-2/b29-13-,30-14-,31-13-,32-15-,33-16-,34-16-,35-14-,36-15-;
Formel vægt 884.28