missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytidine-5'-Triphosphate Disodium Salt Dihydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
1 g
Packaging:
Plastic bottle
Specifications
Physical Form | Powder |
Quantity | 100 mg |
Formula Weight | 527.13 (anhydrous) |
Packaging | Plastic bottle |
Chemical Name or Material | Cytidine to 5 '-Triphosphate Disodium Salt Dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'