missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Cytidine-5'-triphosphate disodium salt, 97%
Click to view available options
Quantity:
500 mg
1 g
2 g
Description
Cytidine-5'-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.
Notes
Incompatible with strong oxidizing agents.
Cytidine-5′-triphosphate disodium salt is a cation-permeable ligand gated ion channel (P2X purinergic receptor) agonist. It is also used as an organic intermediate, pharmaceutical intermediate, food additive and flavoring agent.
Solubility
Soluble in water. Insoluble in ethanol, chloroform and ether.
Notes
Incompatible with strong oxidizing agents.
Specifications
Color | White |
Flash Point | 113°C (235°F) |
Quantity | 500 mg |
Solubility Information | Soluble in water. Insoluble in ethanol,chloroform and ether. |
Formula Weight | 527.12 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Cytidine-5'-triphosphate disodium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'