| InChI formel |
InChI=1S/C14H26N4O11P2/c1-18(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)17-5-4-10(15)16-14(17)21/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H3-,15,16,21,22,23,24,25)/t9-,11-,12-,13-/m1/s1 |
| Synonym |
Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, inner salt, Choline, hydroxide, 5'-ester with cytidine 5'-(trihydrogen pyrophosphate), inner salt (8CI), Cytidine 5'-(trihydrogen diphosphate), P'-[2-(trimethylammonio)ethyl] ester, hydroxide, inner salt, Audes, CDP-choline, Cereb, Choline 5'-cytidine diphosphate, Choline cytidine diphosphate, Citicholine, Citicoline, Citidoline, Citifar, Cognizin, Colite, Corenalin, Cyscholin, Cytidine 5'-(choline diphosphate), Cytidine 5'-(cholinyl pyrophosphate), Cytidine 5'-diphosphate choline, Cytidine 5'-diphosphocholine, Cytidine choline diphosphate, Cytidine diphosphate choline, Cytidine diphosphate choline ester, Cytidine diphosphocholine, Cytidine diphosphorylcholine, Cytidoline, Difosfocin, Emicholine F, Ensign, Haocolin, Hornbest, NSC 122002, Neucolis, Nicholin, Nicolin, Niticolin, Reagin, Recofnan, Recognan, Rexort, Sintoclar, Somazina, Somazine, Suncholin |