missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 2':3'-cyclic Monophosphate Monosodium Salt (>85%), TRC

Product Code. 30363507
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30363507 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363507 Supplier Toronto Research Chemicals Supplier No. TRCC99835525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Cytidine 2':3'-cyclic Monophosphate Monosodium Salt
CAS 15718-51-1
Molekylær formel C9H11N3NaO7P
Mængde 25 mg
Synonym Furo[3,4-d]-1,3,2-dioxaphosphole, Cytidine derivative, Cyclic(2',3')-CMP, Sodium SaltCytidine 2',3'-cyclic Phosphate Sodium SaltNSC 238924Sodium Cytidine-2',3'-cyclophosphate
InChI formel InChI=1S/C9H12N3O7P.Na/c10-5-1-2-12(9(14)11-5)8-7-6(4(3-13)17-8)18-20(15,16)19-7;/h1-2,4,6-8,13H,3H2,(H,15,16)(H2,10,11,14);/q;+1/p-1/t4-,6-,7-,8-;/m1./s1
SMIL [Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@H]3OP(=O)([O-])O[C@@H]23
IUPAC navn sodium;1-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-4-yl]-4-aminopyrimidin-2-one
Molekylvægt (g/mol) 327.16
Formel vægt 327.0232
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.