Få mere at vide
Cystamine-d8 2HCl, CDN
Artikelnummer. 30320522
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320522 0.05 g
30313414 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320522 Leverandør CDN Leverandørnr. CDND79560.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cystamine-D8 Dihydrochloride
CAS 2712126-51-5
Molekylær formel C4 D8 H4 N2 S2 . 2 Cl H
Mængde 0.05 g
Synonym Cystamine-d8 2 HCl, 2,2'-Dithiobis[ethan-1,1,2,2-d4-amine] hydrochloride (1:2), Cystamine-d8 dihydrochloride, Cystamine-D8 dihydrochloride
InChI formel InChI=1 S/C4H12N2S2.2 ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1 H/i1D2,2D2,3D2,4D2;;
SMIL Cl.Cl.[2 H]C([2 H])(N)C([2 H])([2 H])SSC([2 H])([2 H])C([2 H])([2 H])N
IUPAC navn 2-[(2-amino-1,1,2,2-tetradeuterioethyl)disulfanyl]-1,1,2,2-tetradeuterioethanamine;dihydrochloride
Molekylvægt (g/mol) 233.2527
Formel vægt 232.0478 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity