Få mere at vide
Cyproheptadine beta-N-Oxide, Mikromol™
Artikelnummer. 30050060
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050060 25 mg
30051919 100 mg
30461751 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050060 Leverandør Mikromol Leverandørnr. MM0609.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Cyproheptadine beta-N-Oxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyproheptadine beta-N-Oxide
CAS 60251-34-5
Analyt- eller komponentnavne Cyproheptadine beta-N-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C21 H21 N O
Certificeringer/compliance ISO 9001
Synonym Cyproheptadine beta-N-Oxide
InChI formel InChI=1S/C21H21NO/c1-22(23)14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21/h2-11H,12-15H2,1H3
SMIL C[N+]1([O-])CCC(=C2c3ccccc3C=Cc4ccccc24)CC1
IUPAC navn 4-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-1-methyl-1-oxidopiperidin-1-ium
Molekylvægt (g/mol) 303.4
Mængde 25 mg
Formel vægt 303.1623 g/mol