Få mere at vide
Cyprodinil-d5, TRC
Artikelnummer. 30354274
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30354274 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354274 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9890625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyprodinil phenyl-d5
CAS 1773496-67-5
Renhedsgrad noter HPLC
Molekylær formel C14 2H5 H10 N3
Mængde 5 mg
Synonym Cyprodinil D5 (phenyl D5)
InChI formel InChI=1S/C14H15N3/c1-10-9-13(11-7-8-11)17-14(15-10)16-12-5-3-2-4-6-12/h2-6,9,11H,7-8H2,1H3,(H,15,16,17)/i2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(Nc2nc(C)cc(n2)C3CC3)c([2H])c1[2H]
IUPAC navn 4-cyclopropyl-6-methyl-N-(2,3,4,5,6-pentadeuteriophenyl)pyrimidin-2-amine
Molekylvægt (g/mol) 230.32
Formel vægt 230.158
Procent renhed >95