Få mere at vide
Cyprodinil-13C6, TRC
Artikelnummer. 30136653
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136653 5 mg
30135681 10 mg
30007643 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136653 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9890615MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1773496-63-1
Molekylær formel 13C6 C8 H15 N3
Molekylvægt (g/mol) 231.24
Synonym Cyprodinil 13C6 (phenyl 13C6)
IUPAC navn N-((1,2,3,4,5,6-^{13}C_{6})cyclohexatrienyl)-4-cyclopropyl-6-methylpyrimidin-2-amine
SMIL Cc1cc(nc(N[13c]2[13cH][13cH][13cH][13cH][13cH]2)n1)C3CC3
Tekniske data
Kemisk navn eller materiale Cyprodinil 13C6 (phenyl 13C6)
Mængde 5 mg
InChI formel InChI=1S/C14H15N3/c1-10-9-13(11-7-8-11)17-14(15-10)16-12-5-3-2-4-6-12/h2-6,9,11H,7-8H2,1H3,(H,15,16,17)/i2+1,3+1,4+1,5+1,6+1,12+1
Formel vægt 231.1467