Få mere at vide
Cypermethrin (Phenoxy-d5) Isomeric Mixture, TRC
Artikelnummer. 30240963
Skift visning
Mængde:
1 mg
2.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240963 2.5 mg
30240821 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240963 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9890022.5MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2140327-50-8
Molekylær formel C22 D5 H14 Cl2 N O3
Molekylvægt (g/mol) 421.328
Synonym 3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarboxylic Acid-d5 Cyano(3-phenoxyphenyl)methyl Ester, NRDC-149-d5, FMC-30980-d5, PP-383-d5, Ammo-d5, Arrivo-d5, Barricade-d5, Basathrin-d5, Cymbush-d5, Demon-d5, Flectron-d5, Ripcord-d5;
IUPAC navn [cyano-[3-(2,3,4,5,6-pentadeuteriophenoxy)phenyl]methyl] 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
SMIL [2H]c1c([2H])c([2H])c(Oc2cccc(c2)C(OC(=O)C3C(C=C(Cl)Cl)C3(C)C)C#N)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale Cypermethrin (Phenoxy-d5) Isomeric Mixture
Analyt- eller komponentnavne Cypermethrin (Phenoxy-d5) Isomeric Mixture
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/i3D,4D,5D,8D,9D
Løsningstype Neat
Industry Type Environmental Testing
Mængde 2.5 mg
Formel vægt 420.106
Procent renhed >95%