missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP20A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551248
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CYP20A1 protein.
CYP20A1 Polyclonal antibody specifically detects CYP20A1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CYP20A1 | |
Polyclonal | |
Unconjugated | |
CYP20A1 | |
A930011N14Rik; Cyp20a1; Cypm; CYP-M; cytochrome P450 20A1; cytochrome P450 family 20 subfamily A member 1; cytochrome P450 monooxygenase; cytochrome P450, 20a1; cytochrome P450, family 20, subfamily A, polypeptide 1; UNQ667/PRO1301 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57404, 77951 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q6UW02, Q8BKE6 | |
CYP20A1 | |
Fusion protein of human CYP20A1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |