Få mere at vide
Cymoxanil-d3 (methoxy-d3), TRC
Artikelnummer. 30350984
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30350984 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350984 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9889962.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cymoxanil methoxy-d3
CAS 2140803-92-3
Molekylær formel C7 2H3 H7 N4 O3
Mængde 2.5 mg
Synonym Cymoxanil D3 (methoxy D3)
InChI formel InChI=1S/C7H10N4O3/c1-3-9-7(13)10-6(12)5(4-8)11-14-2/h3H2,1-2H3,(H2,9,10,12,13)/b11-5+/i2D3
SMIL [2H]C([2H])([2H])O\N=C(/C#N)\C(=O)NC(=O)NCC
IUPAC navn (1E)-2-(ethylcarbamoylamino)-2-oxo-N-(trideuteriomethoxy)ethanimidoyl cyanide
Molekylvægt (g/mol) 201.2
Formel vægt 201.0941