Få mere at vide
λ-Cyhalothrin, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | lambda-Cyhalothrin |
| CAS | 91465-08-6 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C23 H19 Cl F3 N O3 |
| Mængde | 1 g |
| Synonym | Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, cyano(3-phenoxyphenyl)methyl ester, [1α(S*),3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-rel- (9CI), A 12871A, Ambush Super, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (1R)-cyano(3-phenoxyphenyl)methyl ester, (1S,3S)-, Cyhalothrin K, Demand, Demand Pestab, Grenade ER, Hallmark, ICI-A 0321, ICON Maxx, Icon, Karate, Karate 1E, Karate 2.5EC, Karate K, Karate King, Karate Z, Karate Zeon, Karate Zeon 050CS, Karate Zeon 100CS, Kung Fu, Lamdex, Matador, Matador (insecticide), Molniya, NUP 05077, PP 321, REEVA 5, Scimitar, Spectracide Trazicide, Spectracide Triazicide Soil and Turf Insect Killer, Taiga Z, Warrior, Warrior II, Warrior T, Warrior Zeon, lambda-Cyhalothrin, λ-Cyhalothrin |
| InChI formel | InChI=1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-/t17-,18+,20-/m1/s1 |
| SMIL | CC1(C)[C@H](\C=C(/Cl)\C(F)(F)F)[C@@H]1C(=O)O[C@@H](C#N)c2cccc(Oc3ccccc3)c2 |
| IUPAC navn | [(R)-cyano-(3-phenoxyphenyl)methyl] (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate |
| Molekylvægt (g/mol) | 449.85 |
| Vis mere |