Få mere at vide
Cycrimine, TRC
Artikelnummer. 30356677
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356677 100 mg
30352213 500 mg
30351803 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30356677 Leverandør Toronto Research Chemicals Leverandørnr. TRCC991445100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 77-39-4
Molekylær formel C19H29NO
Molekylvægt (g/mol) 287.44
Synonym alpha-Cyclopentyl-alpha-phenyl-1-piperidinepropanol, 1-Phenyl-1-cyclopentyl-3-piperidino-1-propanol, Pagitane
IUPAC navn 1-cyclopentyl-1-phenyl-3-piperidin-1-ylpropan-1-ol
SMIL OC(CCN1CCCCC1)(C2CCCC2)c3ccccc3
Tekniske data
Kemisk navn eller materiale Cycrimine
Mængde 100 mg
InChI formel InChI=1S/C19H29NO/c21-19(18-11-5-6-12-18,17-9-3-1-4-10-17)13-16-20-14-7-2-8-15-20/h1,3-4,9-10,18,21H,2,5-8,11-16H2