Få mere at vide
Cycotiamine, TRC
Artikelnummer. 30366421
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366421 1 g
30362368 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366421 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9889701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6092-18-8
Molekylær formel C13 H16 N4 O3 S
Molekylvægt (g/mol) 308.36
Synonym N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[1-(2-oxo-1,3-oxathian-4-ylidene)ethyl]formamide, CCT, Carbothiamine, Cometamine, Commetamin, Cyclocarbothiamine, Cyclothiamine, Cyclotiamine, Cycothiamine, Thiocarbonic Acid Cyclic O,S-ester with N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-(4-hydroxy-2-mercapto-1-methyl-1-butenyl)formamide
IUPAC navn N-[(4-amino-2-methylpyrimidin-5-yl)methyl]-N-[(1E)-1-(2-oxo-1,3-oxathian-4-ylidene)ethyl]formamide
SMIL C\C(=C/1\CCOC(=O)S1)\N(Cc2cnc(C)nc2N)C=O
Tekniske data
Kemisk navn eller materiale Cycotiamine
Mængde 1 g
InChI formel InChI=1S/C13H16N4O3S/c1-8(11-3-4-20-13(19)21-11)17(7-18)6-10-5-15-9(2)16-12(10)14/h5,7H,3-4,6H2,1-2H3,(H2,14,15,16)/b11-8+
Formel vægt 308.0943