Få mere at vide
Cyclosporin AM 1 (>80%), TRC
Artikelnummer. 30366141
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366141 10 mg
30350428 5 mg
30355419 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366141 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98891510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 89270-28-0
Molekylær formel C62 H111 N11 O13
Molekylvægt (g/mol) 1218.61
Synonym Cyclosporin A, 6-[(2S,3R,4R,6E)-3,8-dihydroxy-4-methyl-2-(methylamino)-6-octenoic acid]-, Cyclosporin A, 6-[(3R,4R,6E)-6,7-didehydro-3,8-dihydroxy-N,4-dimethyl-L-2-aminooctanoic acid]-, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., 6-[(2S,3R,4R,6E)-3,8-Dihydroxy-4-methyl-2-(methylamino)-6-octenoic acid]cyclosporin A, 7: PN: US6686454 SEQID: 7 claimed protein, AM 1, AM 1 (peptide), Cyclosporin A metabolite 17, Cyclosporin AM 1, Cyclosporin metabolite 17, M 17, OL 17
IUPAC navn (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-33-[(E,1R,2R)-1,6-dihydroxy-2-methylhex-4-enyl]-30-ethyl-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
SMIL CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\CO)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
Tekniske data
Kemisk navn eller materiale Cyclosporin AM 1
Mængde 10 mg
InChI formel InChI=1S/C62H111N11O13/c1-24-43-58(82)67(17)33-48(75)68(18)44(29-34(2)3)55(79)66-49(38(10)11)61(85)69(19)45(30-35(4)5)54(78)63-41(15)53(77)64-42(16)57(81)70(20)46(31-36(6)7)59(83)71(21)47(32-37(8)9)60(84)72(22)50(39(12)13)62(86)73(23)51(56(80)65-43)52(76)40(14)27-25-26-28-74/h25-26,34-47,49-52,74,76H,24,27-33H2,1-23H3,(H,63,78)(H,64,77)(H,65,80)(H,66,79)/b26-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1
Formel vægt 1217.8363