Få mere at vide
MedChemExpress Cyclosporin A-Derivative 1 Free base
Artikelnummer. 16402088
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16402088 5 mg
1 options
Artikelnummer. 16402088 Leverandør MedChemExpress Leverandørnr. HYP1355A
MedChemExpress Cyclosporin A-Derivative 1 (Free base) is a crystalline intermediate derived from the opening of cyclosporin A extracted from patent WO 2013167703 A1. Cyclosporin A is an immunosuppressive agent which can bind to the cyclophilin and inhibit calcineurin.
Tekniske data
Renhed 99.29%
CAS 286852-20-8
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molekylvægt (g/mol) 1276.69 g/mol
Produkttype Peptides