Få mere at vide
Cycloserine Diketopiperazine, TRC
Artikelnummer. 30364312
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364312 250 mg
30353477 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364312 Leverandør Toronto Research Chemicals Leverandørnr. TRCC988810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1204-99-5
Molekylær formel C6H12N4O4
Molekylvægt (g/mol) 204.18
Synonym 3,6-Bis[(aminooxy)methyl]-2,5-piperazinedione, 2,5-Bis(aminohydroxymethyl)-2,6- diketopiperazine, 2,5-Bis(aminooxymethyl)-3,6-dioxopiperazine, Cycloserine Diketopiperazine, NSC 119130
IUPAC navn 3,6-bis(aminooxymethyl)piperazine-2,5-dione
SMIL C(C1C(=O)NC(C(=O)N1)CON)ON
Tekniske data
Kemisk navn eller materiale Cycloserine Diketopiperazine
InChI formel InChI=1S/C6H12N4O4/c7-13-1-3-5(11)10-4(2-14-8)6(12)9-3/h3-4H,1-2,7-8H2,(H,9,12)(H,10,11)
Mængde 250 mg
Formel vægt 204.0859