Få mere at vide
Cyclopropylmethanol-d4, TRC
Artikelnummer. 30361655
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30361655 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361655 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98860825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropylmethanol-d4
CAS 91314-18-0
Molekylær formel C4 D4 H4 O
Mængde 25 mg
Synonym Cyclopropane-2,2,3,3-d4-methanol (7CI), (2,2,3,3-Tetradeuteriocyclopropyl)methanol, Cyclopropylmethanol-d4, Cyclopropyl-2,2,3,3-d4-methyl Alcohol, Cyclopropylmethanol-D4 (cyclopropyl-2,2,3,3-D4), Cyclopropylmethanol-2',2',3',3'-D4
InChI formel InChI=1S/C4H8O/c5-3-4-1-2-4/h4-5H,1-3H2/i1D2,2D2
SMIL [2H]C1([2H])C(CO)C1([2H])[2H]
IUPAC navn (2,2,3,3-tetradeuteriocyclopropyl)methanol
Molekylvægt (g/mol) 76.13
Formel vægt 76.083