Få mere at vide
Cyclopropylhydrazine Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cyclopropylhydrazine Dihydrochloride |
| CAS | 1374652-23-9 |
| Molekylær formel | C3H8N2 . 2 HCl |
| Mængde | 100 mg |
| Synonym | Hydrazine, cyclopropyl-, hydrochloride (1:2) (ACI), 1-Cyclopropylhydrazine 2HCl, Cyclopropylhydrazine dihydrochloride |
| InChI formel | InChI=1S/C3H8N2.2ClH/c4-5-3-1-2-3;;/h3,5H,1-2,4H2;2*1H |
| SMIL | Cl.Cl.NNC1CC1 |
| IUPAC navn | cyclopropylhydrazine;dihydrochloride |
| Molekylvægt (g/mol) | 145.03 |
| Formel vægt | 144.0221 |