Få mere at vide
Cyclopropyl-d5-amine, TRC
Artikelnummer. 30359615
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30359615 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359615 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98914325MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropyl-d5-amine
CAS 153557-95-0
Molekylær formel C3 D5 H2 N
Mængde 25 mg
Synonym Cyclopropan-d5-amine (9CI), 1,2,2,3,3-Pentadeuteriocyclopropan-1-amine, Cyclopropylamine-D5, Aminocyclopropane-1,2,2,3,3-d5
InChI formel InChI=1S/C3H7N/c4-3-1-2-3/h3H,1-2,4H2/i1D2,2D2,3D
SMIL [2H]C1([2H])C([2H])([2H])C1([2H])N
IUPAC navn 1,2,2,3,3-pentadeuteriocyclopropan-1-amine
Molekylvægt (g/mol) 62.125
Formel vægt 62.089