Få mere at vide
Cyclopropyl-d5-amine, CDN
Artikelnummer. 30320776
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320776 0.25 g -
30314481 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320776 Leverandør CDN Leverandørnr. CDND16590.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropyl-d5-amine
CAS 153557-95-0
Molekylær formel C3 D5 H2 N
Mængde 0.25 g
Synonym Cyclopropan-d5-amine (9 CI), 1,2,2,3,3-Pentadeuteriocyclopropan-1-amine, Cyclopropylamine-D5, Aminocyclopropane-1,2,2,3,3-d5
InChI formel InChI=1 S/C3H7N/c4-3-1-2-3/h3H,1-2,4H2/i1D2,2D2,3 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C1([2 H])N
IUPAC navn 1,2,2,3,3-pentadeuteriocyclopropan-1-amine
Molekylvægt (g/mol) 62.125
Formel vægt 62.089 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity