Få mere at vide
Cyclopropane-d5-carboxylic Acid, CDN
Artikelnummer. 30309893
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309893 0.25 g
30319969 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309893 Leverandør CDN Leverandørnr. CDND66990.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropanecarboxylic Acid-D5
CAS 1219804-09-7
Molekylær formel C4 D5 H O2
Mængde 0.25 g
Synonym Cyclopropane-1,2,2,3,3-d5-carboxylic acid (ACI), Cyclopropanecarboxylic acid-1,2,2,3,3-d5, Cyclopropane-d5-carboxylic acid, Cyclopropanecarboxylic acid-D5
InChI formel InChI=1 S/C4H6O2/c5-4(6)3-1-2-3/h3H,1-2H2,(H,5,6)/i1D2,2D2,3 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C1([2 H])C(=O)O
IUPAC navn 1,2,2,3,3-pentadeuteriocyclopropane-1-carboxylic acid
Molekylvægt (g/mol) 91.1201
Formel vægt 91.0682 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity