Få mere at vide
Cyclopropane-2,2,3,3-d4-carboxylic Acid, CDN
Artikelnummer. 30314485
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314485 0.5 g 0.5g
30323591 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314485 Leverandør CDN Leverandørnr. CDND63900.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropanecarboxylic Acid-2,2,3,3-D4
CAS 89924-82-3
Molekylær formel C4 D4 H2 O2
Mængde 0.5 g
Synonym Cyclopropanecarboxylic-2,2,3,3-d4 acid (7 CI, ACI), Cyclopropane-2,2,3,3-d4-carboxylic acid, Cyclopropanecarboxylic acid-2,2,3,3-D4
InChI formel InChI=1 S/C4H6O2/c5-4(6)3-1-2-3/h3H,1-2H2,(H,5,6)/i1D2,2D2
SMIL [2 H]C1([2 H])C(C(=O)O)C1([2 H])[2 H]
IUPAC navn 2,2,3,3-tetradeuteriocyclopropane-1-carboxylic acid
Molekylvægt (g/mol) 90.1139
Formel vægt 90.0619 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity