Få mere at vide
Cyclopropane-1-d1-carboxylic Acid, CDN
Artikelnummer. 30311523
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311523 1 g
30320802 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311523 Leverandør CDN Leverandørnr. CDND14011G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopropanecarboxylic Acid-1-D
CAS 19136-94-8
Molekylær formel C4 D H5 O2
Mængde 1 g
Synonym Cyclopropane-1-d-carboxylic acid (8 CI, 9 CI), Cyclopropanecarboxylic acid-1-D
InChI formel InChI=1 S/C4H6O2/c5-4(6)3-1-2-3/h3H,1-2H2,(H,5,6)/i3D
SMIL [2 H]C1(CC1)C(=O)O
IUPAC navn 1-deuteriocyclopropane-1-carboxylic acid
Molekylvægt (g/mol) 87.0954
Formel vægt 87.0431 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity