Få mere at vide
Cyclopentylmethylamine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cyclopentylmethylamine |
| CAS | 2439-56-7 |
| Molekylær formel | C6H13N |
| Mængde | 500 mg |
| Synonym | N-Methylcyclopentanamine, (Methylamino)cyclopentane, Methyl(cyclopentyl)amine, N-Cyclopentyl-N-methylamine, N-Methylcyclopentylamine |
| InChI formel | InChI=1S/C6H13N/c1-7-6-4-2-3-5-6/h6-7H,2-5H2,1H3 |
| SMIL | CNC1CCCC1 |
| IUPAC navn | N-methylcyclopentanamine |
| Molekylvægt (g/mol) | 99.17 |
| Formel vægt | 99.1048 |