Få mere at vide
Cyclopentylalbendazole-sulfoxide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Cyclopentylalbendazole-sulfoxide |
| CAS | 131454-43-8 |
| Analyt- eller komponentnavne | Cyclopentylalbendazole-sulfoxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H17 N3 O3 S |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Cyclopentylalbendazole-sulfoxide, Carbamic acid, [5-(cyclopentylsulfinyl)-1H-benzimidazol-2-yl]-, methyl ester |
| InChI formel | InChI=1S/C14H17N3O3S/c1-20-14(18)17-13-15-11-7-6-10(8-12(11)16-13)21(19)9-4-2-3-5-9/h6-9H,2-5H2,1H3,(H2,15,16,17,18) |
| SMIL | COC(=O)Nc1nc2cc(ccc2[nH]1)S(=O)C3CCCC3 |
| Vis mere |