missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl thiocyanate, 95%, Thermo Scientific™

Catalog Number p-7031229
Click to view available options
Quantity:
500 mg
2 g
This item is not returnable. View return policy
5263-57-0
C6H9NS
127.21
MFCD11100558
ZTZNTHBAUFZQGQ-UHFFFAOYSA-N
cyclopentanethiocyanate, cyclopentylsulfanyl formonitrile, cyclopentylrhodanid
12376019
cyclopentyl thiocyanate
N#CSC1CCCC1
This item is not returnable. View return policy
5263-57-0
C6H9NS
127.21
MFCD11100558
ZTZNTHBAUFZQGQ-UHFFFAOYSA-N
cyclopentanethiocyanate, cyclopentylsulfanyl formonitrile, cyclopentylrhodanid
12376019
cyclopentyl thiocyanate
N#CSC1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 84°C (8 mmHg)
Quantity 500 mg
Formula Weight 127.21
Percent Purity 95%
Chemical Name or Material Cyclopentyl thiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'