Få mere at vide
Cyclopentanone-d8, CDN
Artikelnummer. 30317530
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317530 0.25 g
30315388 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317530 Leverandør CDN Leverandørnr. CDND01010.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopentanone-D8
CAS 4477-17-2
Molekylær formel C5 D8 O
Mængde 0.25 g
Synonym Cyclopentanone-d8 (7 CI, 8 CI, 9 CI), Octadeuterocyclopentanone, Cyclopentanone-2,2,3,3,4,4,5,5-d8, Cyclopentanone-D8
InChI formel InChI=1 S/C5H8O/c6-5-3-1-2-4-5/h1-4H2/i1D2,2D2,3D2,4D2
SMIL [2 H]C1([2 H])C(=O)C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 2,2,3,3,4,4,5,5-octadeuteriocyclopentan-1-one
Molekylvægt (g/mol) 92.1657
Formel vægt 92.1077 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity