Få mere at vide
Cyclopentanone-3,3,4,4-d4, CDN
Artikelnummer. 30326081
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326081 0.5 g 0.5g
30315640 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326081 Leverandør CDN Leverandørnr. CDND51520.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3,4,4-tetradeuteriocyclopentan-1-one
CAS 36219-23-5
Molekylær formel C5 2H4 H4 O
Mængde 0.5 g
InChI formel InChI=1 S/C5H8O/c6-5-3-1-2-4-5/h1-4H2/i1D2,2D2
SMIL [2 H]C1([2 H])CC(=O)CC1([2 H])[2 H]
IUPAC navn 3,3,4,4-tetradeuteriocyclopentan-1-one
Molekylvægt (g/mol) 88.141
Formel vægt 88.083 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity