Learn More
Cyclopentane, Sigma-Aldrich™
Product Code. 15840611
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15840611 1 L 1 L
15830611 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15840611 Supplier Sigma Aldrich Supplier No. 8187691000
 Restricted Item
Forventet forsendelse: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 287-92-3
Specifications
Melting Point -93.3°C
Density 0.75 g/cm3 at 20°C
Boiling Point 49°C
MDL Number MFCD00001356
Quantity 1 L
Synonym Cyclopentane, Cyclopentane
Molecular Weight (g/mol) 70.13
Grade Research