Få mere at vide
Cyclopentane-d10, TRC
Artikelnummer. 30417374
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30417374 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417374 Leverandør Toronto Research Chemicals Leverandørnr. TRCP84083410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopentane-d10
CAS 7426-92-8
Molekylær formel C5 D10
Mængde 10 mg
Synonym Cyclopentane-d10 (6CI','7CI','8CI','9CI','ACI), Perdeuteriocyclopentane, Cyclopentane-D10
InChI formel InChI=1S/C5H10/c1-2-4-5-3-1/h1-5H2/i1D2,2D2,3D2,4D2,5D2
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5-decadeuteriocyclopentane
Molekylvægt (g/mol) 80.195
Formel vægt 80.141