Få mere at vide
Cyclopentane-d10, CDN
Artikelnummer. 30313139
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313139 1 g
30321346 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313139 Leverandør CDN Leverandørnr. CDND02361G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclopentane-d10
CAS 7426-92-8
Molekylær formel C5 D10
Mængde 1 g
Synonym Cyclopentane-d10 (6 CI, 7 CI, 8 CI, 9 CI, ACI), Perdeuteriocyclopentane, Cyclopentane-D10
InChI formel InChI=1 S/C5H10/c1-2-4-5-3-1/h1-5H2/i1D2,2D2,3D2,4D2,5D2
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5-decadeuteriocyclopentane
Molekylvægt (g/mol) 80.195
Formel vægt 80.141 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity