Få mere at vide
Cyclooctane-d16, CDN
Artikelnummer. 30315528
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30315528 1 g -
30308962 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315528 Leverandør CDN Leverandørnr. CDND54711G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclooctane-d16
CAS 92204-03-0
Molekylær formel C8 D16
Mængde 1 g
Synonym Cyclooctane-d16 (7 CI, 9 CI), Cyclooctane-D16
InChI formel InChI=1 S/C8H16/c1-2-4-6-8-7-5-3-1/h1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecadeuteriocyclooctane
Molekylvægt (g/mol) 128.3112
Formel vægt 128.2256 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity