Learn More
RETSCH Cyclone Holder
Use this holder with Retsch™ AS 200 jet for sieve cuts of powdered materials which require efficient dispersion and desagglomeration. Retsch™ Cyclone Holders make the collection of two fractions possible with the use of the cyclone.
Brand: RETSCH 32.935.0008
Additional Details : Weight : 4.50000kg
Description
- For use with Air Jet Sieving Machine AS 200 jet
- Cyclone with holder and collecting receptacle
- Includes: 250mL and 500mL sample bottle
Specifications
Cyclone Holder | |
Air Jet Sieving Machine AS 200 jet | |
1 Ea. |