Learn More
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Catalog Number p-96817
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
827-52-1
C12H16
160.26
MFCD00001451
IGARGHRYKHJQSM-UHFFFAOYSA-N
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
13229
cyclohexylbenzene
C1CCC(CC1)C1=CC=CC=C1
827-52-1
C12H16
160.26
MFCD00001451
IGARGHRYKHJQSM-UHFFFAOYSA-N
phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
13229
cyclohexylbenzene
C1CCC(CC1)C1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 5°C
Color Colorless
Density 0.9500g/mL
Boiling Point 238°C to 240°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H11
Refractive Index 1.5250 to 1.527
Quantity 100 mL
Beilstein 05, 503
Specific Gravity 0.95
Solubility Information Solubility in water: insoluble
Formula Weight 160.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexylbenzene, 98%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.