Få mere at vide
Cyclohexylacetaldehyde (~90%), TRC
Artikelnummer. 30354602
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354602 50 mg
30352040 100 mg
30349320 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30354602 Leverandør Toronto Research Chemicals Leverandørnr. TRCC99121050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5664-21-1
Molekylær formel C8H14O
Molekylvægt (g/mol) 126.2
Synonym 2-Cyclohexylacetaldehyde, Cyclohexylmethyl carboxaldehyde, NSC 244934
IUPAC navn 2-cyclohexylacetaldehyde
SMIL O=CCC1CCCCC1
Tekniske data
Kemisk navn eller materiale Cyclohexylacetaldehyde
Mængde 50 mg
InChI formel InChI=1S/C8H14O/c9-7-6-8-4-2-1-3-5-8/h7-8H,1-6H2
Formel vægt 126.1045