missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl methylphosphonate, 99%, Thermo Scientific Chemicals

Catalog Number 15471987
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
7040-52-0
C8H17O3P
192.195
MFCD08064211
UXITUBFQUAHYSL-UHFFFAOYSA-N
cyclohexyl methyl methylphosphonate, phosphonic acid, methyl-, cyclohexyl methyl ester, cyclohexyl methyl methyl-phosphonate, methoxy methyl phosphoryl oxycyclohexane, methylphosphonic acid methylcyclohexyl ester
574009
[methoxy(methyl)phosphoryl]oxycyclohexane
COP(=O)(C)OC1CCCCC1
This item is not returnable. View return policy
7040-52-0
C8H17O3P
192.195
MFCD08064211
UXITUBFQUAHYSL-UHFFFAOYSA-N
cyclohexyl methyl methylphosphonate, phosphonic acid, methyl-, cyclohexyl methyl ester, cyclohexyl methyl methyl-phosphonate, methoxy methyl phosphoryl oxycyclohexane, methylphosphonic acid methylcyclohexyl ester
574009
[methoxy(methyl)phosphoryl]oxycyclohexane
COP(=O)(C)OC1CCCCC1

Cyclohexyl methyl methylphosphonate or phosphonic acid is used as a micro-nano electronic materials have extended self-assembled monolayers (SAMs) beyond conventional gold/thiol systems. To expand the choice of the substrate used for the preparation of SAMs, the chemical functionalities in film-forming molecules can be altered by introducing phosphate or phosphonate groups. Such polar acidic molecules are capable of interacting with diverse metal-oxide surfaces (e.g., Al2O3, Ta2O5, NbO5, ZrO2 and TiO2) and form films with a similar degree of ordering as for alkyl thiol SAMs on gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 g
Solubility Information It is sparingly soluble in both organic solvents and water,but become more soluble in water when neutralized to phosphonates at neutral to high pH.
Formula Weight 192.09
Percent Purity 99%
Chemical Name or Material Cyclohexyl methyl methylphosphonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'