missing translation for 'onlineSavingsMsg'
Få mere at vide

Cyclohexyl methacrylate, 98%, BLD Pharm™

Artikelnummer. 31022990
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
100 g
500 g
1 kg
Pakningsstørrelse:
100g
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
31022990 25 g 25g
31017311 100 g 100g
31021705 500 g -
31019436 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 31022990 Leverandør BLD Pharm Leverandørnr. BD12276425g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Læg i indkøbskurv
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

(Stabilized with MEHQ)

Specifications

Kemisk navn eller materiale Cyclohexyl methacrylate
CAS 101-43-9
Molekylær formel C10H16O2
MDL nummer MFCD00014292
Mængde 25 g
SMIL CC(C(OC1CCCCC1)=O)=C
Molekylvægt (g/mol) 168.23
Procent renhed 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.