Få mere at vide
Cyclohexyl-d11-methyl Alcohol, CDN
Artikelnummer. 30311607
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311607 0.25 g
30320828 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311607 Leverandør CDN Leverandørnr. CDND59070.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexylmethanol-D11 (cyclohexyl-D11)
CAS 1215077-50-1
Molekylær formel C7 D11 H3 O
Mængde 0.25 g
Synonym Cyclohexane-1,2,2,3,3,4,4,5,5,6,6-d11-methanol (ACI), (Perdeuterocyclohexyl)methanol, Cyclohexyl-d11-methyl Alcohol, Cyclohexylmethanol-d11, Cyclohexylmethanol-D11 (cyclohexyl-D11)
InChI formel InChI=1 S/C7H14O/c8-6-7-4-2-1-3-5-7/h7-8 H,1-6H2/i1D2,2D2,3D2,4D2,5D2,7 D
SMIL [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(CO)C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn (1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)methanol
Molekylvægt (g/mol) 125.2532
Formel vægt 125.1735 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity