Få mere at vide
Cyclohexyl 4-Methylbenzenesulfonate, TRC
Artikelnummer. 30350316
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350316 500 mg
30364097 100 mg
30355181 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30350316 Leverandør Toronto Research Chemicals Leverandørnr. TRCC988453500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 953-91-3
Molekylær formel C13H18O3S
Molekylvægt (g/mol) 254.35
Synonym Cyclohexyl P-Toluenesulfonate
IUPAC navn cyclohexyl 4-methylbenzenesulfonate
SMIL Cc1ccc(cc1)S(=O)(=O)OC2CCCCC2
Tekniske data
Kemisk navn eller materiale Cyclohexyl 4-Methylbenzenesulfonate
Mængde 500 mg
InChI formel InChI=1S/C13H18O3S/c1-11-7-9-13(10-8-11)17(14,15)16-12-5-3-2-4-6-12/h7-10,12H,2-6H2,1H3
Formel vægt 254.0977