Få mere at vide
Cyclohexanone-d6, TRC
Artikelnummer. 30358880
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30358880 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358880 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987982100MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanone-d6
CAS 54513-99-4
Molekylær formel C6 D6 H4 O
Mængde 100 mg
Synonym Anon-d6, Anone-d6, Cyclohexanone-3,3,4,4,5,5-d6, Hexanon-d6, Hytrol O-d6, NSC 5711-d6, Nadone-d6, Pimelic Ketone-d6, Pimelin Ketone-d6, Sextone-d6
InChI formel InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2/i1D2,2D2,3D2
SMIL [2H]C1([2H])CC(=O)CC([2H])([2H])C1([2H])[2H]
IUPAC navn 3,3,4,4,5,5-hexadeuteriocyclohexanone
Molekylvægt (g/mol) 104.18
Formel vægt 104.1108