Få mere at vide
Cyclohexanone-d6, TRC
Artikelnummer. 30368572
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30368572 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368572 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9879821G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanone-d6
CAS 54513-99-4
Molekylær formel C6 D6 H4 O
Mængde 1 g
Synonym Anon-d6, Anone-d6, Cyclohexanone-3,3,4,4,5,5-d6, Hexanon-d6, Hytrol O-d6, NSC 5711-d6, Nadone-d6, Pimelic Ketone-d6, Pimelin Ketone-d6, Sextone-d6
InChI formel InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2/i1D2,2D2,3D2
SMIL [2H]C1([2H])CC(=O)CC([2H])([2H])C1([2H])[2H]
IUPAC navn 3,3,4,4,5,5-hexadeuteriocyclohexanone
Molekylvægt (g/mol) 104.18
Formel vægt 104.1108