Få mere at vide
Cyclohexanol-d12, TRC
Artikelnummer. 30366081
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30366081 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366081 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987961100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanol-d12
CAS 66522-78-9
Molekylær formel C6 2H12 O
Mængde 100 mg
Synonym Cyclohexan-1,2,2,3,3,4,4,5,5,6,6-d11-ol-d (ACI), Cyclohexanol-d12 (9CI), Cyclohexanol-D12
InChI formel InChI=1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6D,7D
SMIL [2H]OC1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6-undecadeuterio-6-deuteriooxycyclohexane
Molekylvægt (g/mol) 112.23
Formel vægt 112.1641