Få mere at vide
Cyclohexanol-d12, CDN
Artikelnummer. 30322015
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30322015 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322015 Leverandør CDN Leverandørnr. CDND02661G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanol-d12
CAS 66522-78-9
Molekylær formel C6 2H12 O
Mængde 1 g
InChI formel InChI=1 S/C6H12O/c7-6-4-2-1-3-5-6/h6-7 H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6 D,7 D
SMIL [2 H]OC1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6-undecadeuterio-6-deuteriooxycyclohexane
Molekylvægt (g/mol) 112.23
Formel vægt 112.1641 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity