Få mere at vide
Cyclohexanol-d11, TRC
Artikelnummer. 30361037
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30361037 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361037 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9879621G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanol-d11
CAS 93131-17-0
Molekylær formel C6 D11 H O
Mængde 1 g
Synonym Cyclohexan-1,2,2,3,3,4,4,5,5,6,6-d11-ol (ACI), Cyclohexan-d11-ol (9CI), Cyclohexanol-D11, Cyclohexanol-d11
InChI formel InChI=1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(O)C([2H])([2H])C1([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexan-1-ol
Molekylvægt (g/mol) 111.2267
Formel vægt 111.1579