Få mere at vide
Cyclohexanecarboxylic-d11 Acid, TRC
Artikelnummer. 30353817
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30353817 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353817 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98818250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanecarboxylic-d11 Acid
CAS 93131-16-9
Molekylær formel C7 D11 H O2
Mængde 50 mg
Synonym Cyclohexane-1,2,2,3,3,4,4,5,5,6,6-d11-carboxylic Acid
InChI formel InChI=1S/C7H12O2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H,8,9)/i1D2,2D2,3D2,4D2,5D2,6D
SMIL [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(C(=O)O)C([2H])([2H])C1([2H])[2H]
IUPAC navn 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 139.24
Formel vægt 139.1528