Få mere at vide
Cyclohexanecarboxylic-1-d1 Acid, CDN
Artikelnummer. 30321661
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321661 0.5 g 0.5g
30316803 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321661 Leverandør CDN Leverandørnr. CDND11870.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Cyclohexanecarboxylic Acid-1-D
CAS 933-37-9
Molekylær formel C7 D H11 O2
Mængde 0.5 g
Synonym Cyclohexane-1-d-carboxylic acid (6 CI, 7 CI, 8 CI, ACI), Cyclohexanecarboxylic Acid-d1, Cyclohexanecarboxylic Acid-1-D
InChI formel InChI=1 S/C7H12O2/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2,(H,8,9)/i6D
SMIL [2 H]C1(CCCCC1)C(=O)O
IUPAC navn 1-deuteriocyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 129.1751
Formel vægt 129.09 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity